Learn More
MSC KaiVac Cleaning Machine, 17.4 gal Solution Tank, 500 PSI
Catalog No. 502839777
Encompass
This mid-size no-touch cleaning machine is designed for hygienic cleaning of various surfaces. It applies an automatically diluted cleaning solution, blasts surfaces and floors with an indoor pressure washer, then vacuums the floor dry, removing soils, moisture, and contaminants and leaving surfaces virtually dry and soil-free.
  • Patented no-touch cleaning process for hygienic cleaning.
  • Automatically diluted cleaning solution applied by an indoor pressure washer.
  • Removes soils and contaminants, leaving floors virtually dry.
  • Equipped with a 17.4 gal solution tank and an 18.1 gal vacuum tank.
  • Features a powerful 500 psi pump system.
  • Removable engine compartment with black box technology.
  • HEPA filtered exhaust and 300 mph HEPA blow feature.
  • Operates on 110 volt - 60 Hz with a 50 ft GFCI electric cord.
Catalog No. 50-283-9777 Supplier MSC Supplier No. 78889847
Add to Cart
Add to Cart
Request a Quote
This mid-size no-touch cleaning machine is designed for hygienic cleaning of various surfaces. It applies an automatically diluted cleaning solution, blasts surfaces and floors with an indoor pressure washer, then vacuums the floor dry, removing soils, moisture, and contaminants and leaving surfaces virtually dry and soil-free.
  • Patented no-touch cleaning process for hygienic cleaning.
  • Automatically diluted cleaning solution applied by an indoor pressure washer.
  • Removes soils and contaminants, leaving floors virtually dry.
  • Equipped with a 17.4 gal solution tank and an 18.1 gal vacuum tank.
  • Features a powerful 500 psi pump system.
  • Removable engine compartment with black box technology.
  • HEPA filtered exhaust and 300 mph HEPA blow feature.
  • Operates on 110 volt - 60 Hz with a 50 ft GFCI electric cord.