Learn More
Medchemexpress LLC Kakkanin | 63770-91-2 | 578.52 | 1 MG

Supplier: Medchemexpress LLC HYN93751MG
Kakkanin is a compound derived from the roots of O. henryi, commonly used in research, particularly for studies related to anti-inflammatory effects.
- Stable under recommended storage conditions
- Optimal storage at 4°C, sealed and away from moisture
- For storage in solvent, -80°C for up to 6 months or -20°C for 1 month is recommended
- Can be shipped at room temperature for durations less than 2 weeks
- Identified as a solid form
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.