Learn More
Bachem KALA Amphipathic Peptide
SDP
Supplier:  Bachem 4030798.0500
Encompass_Preferred
KALA Amphipathic Peptide , H-Trp-Glu-Ala-Lys-Leu-Ala-Lys-Ala-Leu-Ala-Lys-Ala-Leu-Ala-Lys-His-Leu-Ala-Lys-Ala-Leu-Ala-Lys-Ala-Leu-Lys-Ala-Cys-Glu-Ala-OH, 187987-64-0, C144H248N40O35S, Mr 3131.87, 0.5mg. KALA amphipathic peptide, a low-molecular weight cationic amphipathic peptide was able to bind to DNA, to destabilize membranes and to mediate transfection of plasmid DNA in various cell lines. Thus, KALA provides a starting point for a family of peptides that incorporate other functions to improve DNA delivery systems.
Catalog No. 50-194-6814