Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

InBios International Inc. Kalazar Detect™ Rapid Test
SDP

Catalog No. 22277755
Change view
Click to view available options
Quantity:
25 Tests/Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-277-755 25 Tests/Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-277-755 Supplier InBios International Inc. Supplier No. INS025
Add to Cart
Add to Cart
This item is not returnable. View return policy

The Kalazar Detect™ Rapid Test for Visceral Leishmaniasis (VL) is a rapid immunochromatographic strip assay for the qualitative detection of antibodies to members of L. donovani in human serum to aid in the presumptive diagnosis of VL.

  • FDA 510(k) Cleared and CE Marked
  • Relative Sensitivity and Relative Specificity: >90%
  • Long shelf life - two years

Specifications

Content And Storage Room Temp, 20°C to 30°C
Detectable Analytes Antibodies to members of L. donovani
Detection Method Rapid Test
Type Rapid Test
Immunoassay Kit Format ICT (Strip)
Quantity 25 Tests/Kit
Analysis Time 10 min.
Form Ready-to-Use (RTU)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.