Learn More
Cayman Chemical KalIdIn380-389humnpo 10mg
SDP
Catalog No. 502991752
Encompass_Preferred
An endogenous kinin peptide agonist of bradykinin B2 and B1 receptors; binds to bradykinin B2 (IC50 = 0.63 nM) and B1 receptors (Ki = 3.23 nM); levels in interstitial muscle are increased in women with work-related trapezius myalgia
Catalog No. 50-299-1752 Supplier Cayman Chemical Supplier No. 3148110 MG
An endogenous kinin peptide agonist of bradykinin B2 and B1 receptors; binds to bradykinin B2 (IC50 = 0.63 nM) and B1 receptors (Ki = 3.23 nM); levels in interstitial muscle are increased in women with work-related trapezius myalgia