Learn More
Cayman Chemical Kallidin (380-389) (human, porcine, bovine) (trifluoroacetate salt), Purity: ≥98%, 50 mg
SDP
Catalog No. 502992099
Encompass_Preferred
An endogenous kinin peptide agonist of bradykinin B2 and B1 receptors; binds to bradykinin B2 (IC50 = 0.63 nM) and B1 receptors (Ki = 3.23 nM); levels in interstitial muscle are increased in women with work-related trapezius myalgia
Catalog No. 50-299-2099 Supplier Cayman Chemical Supplier No. 3148150 MG
An endogenous kinin peptide agonist of bradykinin B2 and B1 receptors; binds to bradykinin B2 (IC50 = 0.63 nM) and B1 receptors (Ki = 3.23 nM); levels in interstitial muscle are increased in women with work-related trapezius myalgia