Learn More
Medchemexpress LLC Kalii Dehydrographolidi Succinas | 76958-99-1 | 570.67 | 20 MG
SDP
Catalog No. 5000388694
Encompass_Preferred
Kalii Dehydrographolidi Succinas (Potassium dehydroandrographolide succinate) is extracted from the herbal medicine *Andrographis paniculata (Burm f) Nees*. It is widely used for treating viral pneumonia and viral upper respiratory tract infections. Its efficacy is attributed to its immunostimulatory, anti-infective, and anti-inflammatory properties.
  • Extracted from the herbal medicine *Andrographis paniculata (Burm f) Nees*.
  • Widely used for the treatment of viral pneumonia and viral upper respiratory tract infections.
  • Exhibits immunostimulatory, anti-infective, and anti-inflammatory effects.
Catalog No. 50-003-88694 Supplier Medchemexpress LLC Supplier No. HYN0677A20MG
Kalii Dehydrographolidi Succinas (Potassium dehydroandrographolide succinate) is extracted from the herbal medicine *Andrographis paniculata (Burm f) Nees*. It is widely used for treating viral pneumonia and viral upper respiratory tract infections. Its efficacy is attributed to its immunostimulatory, anti-infective, and anti-inflammatory properties.
  • Extracted from the herbal medicine *Andrographis paniculata (Burm f) Nees*.
  • Widely used for the treatment of viral pneumonia and viral upper respiratory tract infections.
  • Exhibits immunostimulatory, anti-infective, and anti-inflammatory effects.