Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Kalii dehydrographolidi succinas | 76958-99-1 | 570.67 | 5 MG
SDP

Catalog No. 5000388509
Encompass_Preferred

Kalii Dehydrographolidi Succinas, extracted from the herbal medicine Andrographis paniculata (Burm f) Nees, is widely used for treating viral pneumonia and viral upper respiratory tract infections due to its immunostimulatory, anti-infective, and anti-inflammatory effects. It is for research use only and not sold to patients.

  • Purity: 98.57%
  • Appearance: Solid
  • Color: White to off-white
  • Structure classification: Terpenoids, Diterpenoids
  • Initial source: Plants (Compositae, Simsia foetida (Cav.) S.F.Blake; Acanthaceae)

Catalog No. 50-003-88509 Supplier Medchemexpress LLC Supplier No. HYN0677A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Kalii Dehydrographolidi Succinas, extracted from the herbal medicine Andrographis paniculata (Burm f) Nees, is widely used for treating viral pneumonia and viral upper respiratory tract infections due to its immunostimulatory, anti-infective, and anti-inflammatory effects. It is for research use only and not sold to patients.

  • Purity: 98.57%
  • Appearance: Solid
  • Color: White to off-white
  • Structure classification: Terpenoids, Diterpenoids
  • Initial source: Plants (Compositae, Simsia foetida (Cav.) S.F.Blake; Acanthaceae)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.