Learn More
Medchemexpress LLC HY-B2233 500mg Medchemexpress, Phosphorylcholine (chloride) CAS:107-73-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB2233500MG
Encompass_Preferred
Medchemexpress, HY-P7381 10ug Kallikrein-11 (Sf9 insect cells-expressed), Human CAS: Recombinant Human Kallikrein-11 (Sf9 insect cells-expressed) is a possible multifunctional protease and efficiently cleaves 'bz-Phe-Arg-4-methylcoumaryl-7-amide', a kallikrein substrate, and weakly cleaves other substrates for kallikrein and trypsin. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0178