Learn More
Medchemexpress LLC Kallikrein-5 Mouse 250ug | 250ug
SDP
Catalog No. 5000187932
Encompass_Preferred
The Kallikrein-5 protein may be involved in desquamation suggesting a role in the process of skin shedding and exfoliation Its association with desquamation suggests that it plays a role in regulating the removal of dead skin cells which is critical for skin homeostasis Kallikrein-5 Protein Mouse (271a a HEK293 His) is the recombinant mouse-derived Kallikrein-5 protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-87932 Supplier Medchemexpress LLC Supplier No. HYP77731A250UG
The Kallikrein-5 protein may be involved in desquamation suggesting a role in the process of skin shedding and exfoliation Its association with desquamation suggests that it plays a role in regulating the removal of dead skin cells which is critical for skin homeostasis Kallikrein-5 Protein Mouse (271a a HEK293 His) is the recombinant mouse-derived Kallikrein-5 protein expressed by HEK293 with C-His labeled tag