Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ProSci Kallikrein 8 Recombinant Protein
SDP

Catalog No. 16593656
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-593-656 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-593-656 Supplier ProSci Supplier No. 913380050
Only null left
Add to Cart
Add to Cart

Recombinant Protein

This recombinant protein can be used for biological assays. For research use only.

Kallikreins are a subgroup of serine proteases having diverse physiological functions. Growing evidence suggests that many Kallikreins are implicated in carcinogenesis and some have potential as novel cancer and other disease biomarkers. This gene is one of the fifteen Kallikrein subfamily members located in a cluster on chromosome 19. Its encoded protein is secreted and may play a role in suppression of tumorigenesis in breast and prostate cancers. Alternate splicing of this gene results in multiple transcript variants encoding the same protein.

Specifications

Accession Number O60259
Gene ID (Entrez) 11202
Molecular Weight (g/mol) 26.04 kDa
Quantity 0.05 mg
Source Human Cells
Common Name kallikrein related peptidase 8
Gene Symbol KLK8
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag C-6 His tag
Sequence Gln29-GLy260
Content And Storage Store at -20°C, stable for 6 months after receipt. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
Purity or Quality Grade >95% as determined by reducing SDS-PAGE. Endotoxin level less than 0.1 ng/μg (1 IEU/μg) as determined by LAL test.
Synonym Kallikrein-8, hK8, Neuropsin, NP, Ovasin, Serine Protease 19, Serine Protease TADG-14, Tumor-Associated Differentially Expressed Gene 14 Protein, KLK8, NRPN, PRSS19, TADG14
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.