Learn More
Athens Research And Technology Inc KALLIKREIN HU PLASMA 50UG
Catalog No. 50489830
Encompass_Preferred
Kallikrein, Human Plasma: 50 UG; Frozen in 20 mM Tris-HCl, pH 7.8, with 100 mM NaCl; MW 86,000-88,000; EC= 1.06; Activity: > =15 U/MG; Purity: > =95% by SDS-PAGE. Native; Prepared and purified from human plasma. Mol. Biology Reagent, Serine Protease
Catalog No. 50-489-830 Supplier Athens Research And Technology Inc Supplier No. 161611011250UG
Kallikrein, Human Plasma: 50 UG; Frozen in 20 mM Tris-HCl, pH 7.8, with 100 mM NaCl; MW 86,000-88,000; EC= 1.06; Activity: > =15 U/MG; Purity: > =95% by SDS-PAGE. Native; Prepared and purified from human plasma. Mol. Biology Reagent, Serine Protease