Learn More
Cusabio Technology LLC kallikrein-related peptidase 5
Catalog No. 5000051238
Encompass_Preferred
This ELISA kit is designed for the quantitative detection of Human Kallikrein-5 (KLK5) in diverse biological samples including serum plasma cell culture supernatants tissue homogenates and saliva Utilizing a sandwich assay format the kit offers a broad detection range of 62 5 pg/mL to 4000 pg/mL with a sensitivity of 15 6 pg/mL ensuring precise measurement of KLK5 concentrations The assay requires only 50-100 L of sample per test and delivers results within 1-5 hours Detection is performed at 450 nm providing reliable and reproducible data for research applications This kit is optimized for accuracy and efficiency making it suitable for studies requiring sensitive quantification of KLK5 in multiple sample matrices
Catalog No. 50-000-51238 Supplier Cusabio Technology LLC Supplier No. CSBEL012456HU
This ELISA kit is designed for the quantitative detection of Human Kallikrein-5 (KLK5) in diverse biological samples including serum plasma cell culture supernatants tissue homogenates and saliva Utilizing a sandwich assay format the kit offers a broad detection range of 62 5 pg/mL to 4000 pg/mL with a sensitivity of 15 6 pg/mL ensuring precise measurement of KLK5 concentrations The assay requires only 50-100 L of sample per test and delivers results within 1-5 hours Detection is performed at 450 nm providing reliable and reproducible data for research applications This kit is optimized for accuracy and efficiency making it suitable for studies requiring sensitive quantification of KLK5 in multiple sample matrices