Learn More
Echelon Biosciences Research Labs Kallikrein Substrate - 10 mg, CAS#: 65147-22-0
SDP
Catalog No. 501909016
Encompass_Preferred
Kallikrein Substrate - 10mg , Z-Phe-Arg-AMC Category/Application: Enzyme Substrates This item is non returnable
This item is not returnable. View return policy
Catalog No. 50-190-9016 Supplier Echelon Biosciences Research Labs Supplier No. 8717510MG
Kallikrein Substrate - 10mg , Z-Phe-Arg-AMC Category/Application: Enzyme Substrates This item is non returnable