Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kalling's Solution (Etchant), Reagents

Catalog No. 01334885
Change view
Click to view available options
Quantity:
120 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-334-885 120 mL
01-334-884 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-334-885 Supplier Reagents Holdings Llc Supplier No. CK017500120A
Add to Cart
Add to Cart

Kalling's Solution (Etchant), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Kalling's Solution (Etchant)
Color Green
CAS 7732-18-5, 67-56-1, 7647-01-0
Boiling Point 80°C
Grade Reagent
Packaging Poly Bottle
Specific Gravity 0.9968
Quantity 120 mL
Sterility Non-sterile
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.