Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kallings Reagent II, Reagents
Kallings Reagent II, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Reagents Holdings Llc CK158000250A
| Quantity | 250 mL |
|---|
Chemical Identifiers
| 7732-18-5, 64-17-5, 7647-01-0 | |
| 18.02 | |
| water |
| H2O | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| O |
Specifications
| Green | |
| H2O | |
| O | |
| 18.02 | |
| Poly Bottle | |
| <1 | |
| Non-sterile | |
| Kallings Reagent II |
| 7732-18-5, 64-17-5, 7647-01-0 | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water | |
| Reagent | |
| 1.015 | |
| 250 mL | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction