Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kallings Reagent II, Reagents

Kallings Reagent II, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

$82.30 - $208.00

Chemical Identifiers

CAS 7732-18-5 , 64-17-5 , 7647-01-0
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
IUPAC Name water
SMILES O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
01-334-879
Accessories View Documents
Reagents Holdings Llc
CK158000100A
100 mL
Each for $82.30
Add to Cart
 
01-334-878
Accessories View Documents
Reagents Holdings Llc
CK158000250A
250 mL
Each for $169.70
Add to Cart
 
01-334-877
Accessories View Documents
Reagents Holdings Llc
CK158000500A
500 mL
Each for $208.00
Add to Cart
 
Description

Description

Specifications

Chemical Identifiers

7732-18-5 , 64-17-5 , 7647-01-0
18.02
water
H2O
XLYOFNOQVPJJNP-UHFFFAOYSA-N
O

Specifications

Green
H2O
O
18.02
Poly Bottle
<1
Liquid
7732-18-5 , 64-17-5 , 7647-01-0
XLYOFNOQVPJJNP-UHFFFAOYSA-N
water
Reagent
1.015
Non-sterile
Kallings Reagent II
Videos
SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.