Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kallings Reagent II, Reagents

Catalog No. 01334879
Change view
Click to view available options
Quantity:
100 mL
250 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-334-879 100 mL
01-334-878 250 mL
01-334-877 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-334-879 Supplier Reagents Holdings Llc Supplier No. CK158000100A
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Kallings Reagent II, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 64-17-5 , 7647-01-0
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
IUPAC Name water
SMILES O

Specifications

Color Green
Grade Reagent
Packaging Poly Bottle
Specific Gravity 1.015
pH <1
Quantity 100 mL
Sterility Non-sterile
Physical Form Liquid
Chemical Name or Material Kallings Reagent II
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.