Learn More
Anton Paar Kalliope Professional
Catalog No. 5000580442
Encompass_Preferred
One-page workflow displays all measurement details in one sight, including input parameters, results, and expert advice.Meets US FDA Regulation 21 CFR Part 11 standards for electronic records, audit trails, and signatures when used in conjunction with theKALLIOPE PROFESSIONAL SOFTWARE MODULE 21 CFR PART 11.Includes license for 3 activations (installations).
Catalog No. 50-005-80442 Supplier Anton Paar Supplier No. 166116
One-page workflow displays all measurement details in one sight, including input parameters, results, and expert advice.Meets US FDA Regulation 21 CFR Part 11 standards for electronic records, audit trails, and signatures when used in conjunction with theKALLIOPE PROFESSIONAL SOFTWARE MODULE 21 CFR PART 11.Includes license for 3 activations (installations).