Learn More
Fine Science Tools KALT SUTURE NEEDLE SIZE 00
SDP
Catalog No. NC9342550
Encompass
Product: Kalt Suture Needles - Size 00/ Size: 00/ Needle Diameter: 0.35mm/ Needle Circle: 3/8/ Needle Arch: 25mm/ Needle Chord: 19.5mm/ Needle Alloy: Stainless Steel/ Sterile: No/ Unit: Dozen
Catalog No. NC9342550 Supplier Fine Science Tools Supplier No. 1205020
Product: Kalt Suture Needles - Size 00/ Size: 00/ Needle Diameter: 0.35mm/ Needle Circle: 3/8/ Needle Arch: 25mm/ Needle Chord: 19.5mm/ Needle Alloy: Stainless Steel/ Sterile: No/ Unit: Dozen