Learn More
Fine Science Tools Kalt Suture Needles, Stainless Steel, Size: 1
SDP
Catalog No. NC9878294
Encompass
Product: Kalt Suture Needles - Size 1/ Size: 1/ Needle Diameter: 0.35mm/ Needle Circle: 3/8/ Needle Arch: 19.8mm/ Needle Chord: 15.5mm/ Needle Alloy: Stainless Steel/ Sterile: No/ Unit: Dozen
Catalog No. NC9878294 Supplier Fine Science Tools Supplier No. 1205001
Product: Kalt Suture Needles - Size 1/ Size: 1/ Needle Diameter: 0.35mm/ Needle Circle: 3/8/ Needle Arch: 19.8mm/ Needle Chord: 15.5mm/ Needle Alloy: Stainless Steel/ Sterile: No/ Unit: Dozen