Learn More
Fine Science Tools Kalt Suture Needles, Stainless Steel, Size: 3
SDP
Catalog No. NC9982303
Encompass
Product: Kalt Suture Needles - Size 3/ Size: 3/ Needle Diameter: 0.35mm/ Needle Circle: 3/8/ Needle Arch: 13.5mm/ Needle Chord: 10.5mm/ Needle Alloy: Stainless Steel/ Sterile: No/ Unit: Dozen
Catalog No. NC9982303 Supplier Fine Science Tools Supplier No. 1205003
Product: Kalt Suture Needles - Size 3/ Size: 3/ Needle Diameter: 0.35mm/ Needle Circle: 3/8/ Needle Arch: 13.5mm/ Needle Chord: 10.5mm/ Needle Alloy: Stainless Steel/ Sterile: No/ Unit: Dozen