Learn More
Medchemexpress LLC HY-112808 10mg Medchemexpress, KAN0438757 CAS:1451255-59-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11280810MG
Encompass_Preferred
Medchemexpress, HY-112808 10mg KAN0438757 CAS:1451255-59-6 KAN0438757 is a potent and selective inhibitor of the metabolic kinase PFKFB3 with an IC50 of 0.19 μM . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9177