Learn More
Selleck Chemical LLC KAN0438757 S0400-5MG
SDP
Catalog No. 5000043496
Encompass_Preferred
KAN0438757 is a potent and selective inhibitor of the metabolic kinase PFKFB3 (6-Phosphofructo-2-Kinase/Fructose-2 6-Biphosphatase 3) with IC50 of 0 19 M and 3 6 M for PFKFB3 and PFKFB4 respectively
Catalog No. 50-000-43496 Supplier Selleck Chemical LLC Supplier No. S04005MG
KAN0438757 is a potent and selective inhibitor of the metabolic kinase PFKFB3 (6-Phosphofructo-2-Kinase/Fructose-2 6-Biphosphatase 3) with IC50 of 0 19 M and 3 6 M for PFKFB3 and PFKFB4 respectively