Learn More
IBI SCIENTIFIC 3P KANAMYCIN 25G
Catalog No. NC0753452
Encompass_Preferred
Kanamycin Sulfate - 25gm Kanamycin Sulfate is used to bind to 70S ribosomal subunit, inhibits translocation, and elicits miscoding bactericidal. Kanamycin Sulfate is derived via microbial fermentation.
Catalog No. NC0753452 Supplier IBI SCIENTIFIC 3P Supplier No. IB02120
Kanamycin Sulfate - 25gm Kanamycin Sulfate is used to bind to 70S ribosomal subunit, inhibits translocation, and elicits miscoding bactericidal. Kanamycin Sulfate is derived via microbial fermentation.