Learn More
Medchemexpress LLC Kanamycin | 59-01-8 | >99.9% | 484.50 | 100 MG
SDP
Catalog No. 5000391274
Encompass_Preferred
Kanamycin (Kanamycin A) is an orally active antibacterial agent that targets gram-negative and gram-positive bacteria. It works by inhibiting translocation and causing misencoding through binding to the 70 S ribosomal subunit.
  • Good inhibitory activity against M. tuberculosis (sensitive and drug-resistant strains) and K. pneumonia.
  • Can be used in studies of tuberculosis and pneumonia.
Catalog No. 50-003-91274 Supplier Medchemexpress LLC Supplier No. HY16566100MG
Kanamycin (Kanamycin A) is an orally active antibacterial agent that targets gram-negative and gram-positive bacteria. It works by inhibiting translocation and causing misencoding through binding to the 70 S ribosomal subunit.
  • Good inhibitory activity against M. tuberculosis (sensitive and drug-resistant strains) and K. pneumonia.
  • Can be used in studies of tuberculosis and pneumonia.