Learn More
Lkt Laboratories Kanamycin A Monosulfate, ≥98%, 1GR
SDP
Supplier:  Lkt Laboratories K00531G
Encompass_Preferred
Aminoglycoside; protein translation inhibitor, mammalian RNA splicing inhibitor. CAS: 25389-94-0. Additional sizes available upon request. Ships from Saint Paul, MN.
Catalog No. 50-153-4914