Learn More
Cusabio Technology LLC Kanamycin KA Elisa Kit
Catalog No. 5000048444
Encompass_Preferred
This ELISA kit detects Kanamycin (KA) in various sample types including milk milk powder and animal tissues such as chicken and pork Designed for high sensitivity it offers a reliable quantitative detection range from 0 5 ppb to 40 5 ppb with a lower detection limit of 0 5 ppb ensuring accurate measurement of trace residues in food safety and veterinary monitoring applications The kit is optimized for ease of use providing consistent performance in screening kanamycin levels to comply with regulatory standards and quality control protocols Its robust design minimizes cross-reactivity and interference making it suitable for laboratories requiring precise analysis of antibiotic residues in complex matrices
Catalog No. 50-000-48444 Supplier Cusabio Technology LLC Supplier No. CSBEFD1027696
This ELISA kit detects Kanamycin (KA) in various sample types including milk milk powder and animal tissues such as chicken and pork Designed for high sensitivity it offers a reliable quantitative detection range from 0 5 ppb to 40 5 ppb with a lower detection limit of 0 5 ppb ensuring accurate measurement of trace residues in food safety and veterinary monitoring applications The kit is optimized for ease of use providing consistent performance in screening kanamycin levels to comply with regulatory standards and quality control protocols Its robust design minimizes cross-reactivity and interference making it suitable for laboratories requiring precise analysis of antibiotic residues in complex matrices