Learn More
Medchemexpress LLC Kanamycin (Kanamycin A) | 59-01-8 | 100.0% | 484.50 | 200 MG
SDP
Catalog No. 5000392174
Encompass_Preferred
Kanamycin (Kanamycin A) is an orally active antibacterial agent effective against gram-negative and gram-positive bacteria. It inhibits translocation and causes misencoding by binding to the 70 S ribosomal subunit. It shows good inhibitory activity against both *M. tuberculosis* (sensitive and drug-resistant) and *K. pneumonia*, and can be used in studies of tuberculosis and pneumonia.
  • Effective against gram-negative and gram-positive bacteria.
  • Inhibits translocation and causes misencoding by binding to the 70 S ribosomal subunit.
  • Shows good inhibitory activity against *M. tuberculosis* (sensitive and drug-resistant).
  • Shows good inhibitory activity against *K. pneumonia*.
  • Suitable for studies of tuberculosis and pneumonia.
Catalog No. 50-003-92174 Supplier Medchemexpress LLC Supplier No. HY16566200MG
Kanamycin (Kanamycin A) is an orally active antibacterial agent effective against gram-negative and gram-positive bacteria. It inhibits translocation and causes misencoding by binding to the 70 S ribosomal subunit. It shows good inhibitory activity against both *M. tuberculosis* (sensitive and drug-resistant) and *K. pneumonia*, and can be used in studies of tuberculosis and pneumonia.
  • Effective against gram-negative and gram-positive bacteria.
  • Inhibits translocation and causes misencoding by binding to the 70 S ribosomal subunit.
  • Shows good inhibitory activity against *M. tuberculosis* (sensitive and drug-resistant).
  • Shows good inhibitory activity against *K. pneumonia*.
  • Suitable for studies of tuberculosis and pneumonia.