Learn More
Alkali Scientific Kanamycin Monosulfate, 5 G
SDP
Catalog No. 5000659733
Encompass_Preferred
Kanamycin Monosulfate is an aminoglycoside antibiotic commonly used for bacterial selection in microbial research and gene expression studies. It works by binding to the 30S subunit of bacterial ribosomes, causing misreading of mRNA and inhibiting protein synthesis. This 5g formulation is typically used in smaller-scale applications where lower concentrations of Kanamycin are required. It is particularly effective against Gram-negative bacteria and is used extensively in molecular biology to select for bacterial strains resistant to this antibiotic, making it an essential reagent in genetic manipulation and cloning experiments.
Catalog No. 50-006-59733 Supplier Alkali Scientific Supplier No. KAN005CP
Kanamycin Monosulfate is an aminoglycoside antibiotic commonly used for bacterial selection in microbial research and gene expression studies. It works by binding to the 30S subunit of bacterial ribosomes, causing misreading of mRNA and inhibiting protein synthesis. This 5g formulation is typically used in smaller-scale applications where lower concentrations of Kanamycin are required. It is particularly effective against Gram-negative bacteria and is used extensively in molecular biology to select for bacterial strains resistant to this antibiotic, making it an essential reagent in genetic manipulation and cloning experiments.