Learn More
NEOGEN Kanamycin Aesculin Azide Agar, 5 kg
SDP
Catalog No. 502015712
Encompass_Preferred
Kanamycin Aesculin Azide Agar is a selective isolation and enumeration medium for enterococci (Lancefield group D streptococci) in food, and is not intended for use in the diagnosis of disease or other conditions in humans.Sodium azide and kanamycin provide the selective inhibition required whilst aesculin and iron salts form an indicator system for the presumptive identification of enterococci. Incubation at 42°C will increase the medium’s selectivity.Formula:Tryptone - 20.0 g/LYeast Extract - 5.0 g/LSodium Chloride - 5.0 g/LSodium Citrate - 1.0 g/LAesculin - 1.0 g/LFerric Ammonium Citrate - 0.5 g/LSodium Azide - 0.15 g/LKanamycin Sulphate - 0.02 g/LAgar- 10.0 g/LFor more information visit https://www.neogen.com/solutions/Microbiology/kanamycin-aesculin-azide-agar
Catalog No. 50-201-5712 Supplier NEOGEN Supplier No. 700004631
Kanamycin Aesculin Azide Agar is a selective isolation and enumeration medium for enterococci (Lancefield group D streptococci) in food, and is not intended for use in the diagnosis of disease or other conditions in humans.Sodium azide and kanamycin provide the selective inhibition required whilst aesculin and iron salts form an indicator system for the presumptive identification of enterococci. Incubation at 42°C will increase the medium’s selectivity.Formula:Tryptone - 20.0 g/LYeast Extract - 5.0 g/LSodium Chloride - 5.0 g/LSodium Citrate - 1.0 g/LAesculin - 1.0 g/LFerric Ammonium Citrate - 0.5 g/LSodium Azide - 0.15 g/LKanamycin Sulphate - 0.02 g/LAgar- 10.0 g/LFor more information visit https://www.neogen.com/solutions/Microbiology/kanamycin-aesculin-azide-agar