Learn More
Ubpbio Kanamycin Sulfate, 100G
Catalog No. 501965188
Encompass_Preferred
Kanamycin sulfate is an aminoglycosidic antibiotic that inhibits the growth of Gram-positive and Gram-negative bacteria and mycoplasmas in cell culture.
Catalog No. 50-196-5188 Supplier Ubpbio Supplier No. P8010100
Kanamycin sulfate is an aminoglycosidic antibiotic that inhibits the growth of Gram-positive and Gram-negative bacteria and mycoplasmas in cell culture.