Learn More
Bioworld Kanamycin Sulfate, 10 g
Supplier: Bioworld 411100003
Kanamycin Sulfate Aminoglycoside with activity against many Gram-negative bacteria. Binds to ribosomal subunit and inhibits protein synthesis. Used as a selective agent for the incorporation of NPT II (Neomycin phosphotransferase) (APH3') gene in plant and animal cells.Biotechnology GradePotency: >750 μg/mgFor in vitro lab use only. Not for human use or consumption.Appearance: SolidBoiling Point : 943.2 °C at 760 mmHgMelting Point: 250 °CRTECS Number: NZ3225030