Learn More
Medchemexpress LLC Kanamycin sulfate | 25389-94-0 | 100.0% | 582.58 | 1 G
SDP
Catalog No. 5000391271
Encompass_Preferred
Kanamycin (Kanamycin A) sulfate is an orally active antibacterial agent effective against gram-negative and gram-positive bacteria. It inhibits translocation and causes misencoding by binding to the 70 S ribosomal subunit. It shows good inhibitory activity against both M. tuberculosis (sensitive and drug-resistant) and K. pneumonia, and is used in studies of tuberculosis and pneumonia.
  • Orally active antibacterial agent.
  • Effective against gram-negative bacteria.
  • Effective against gram-positive bacteria.
  • Inhibits translocation.
  • Causes misencoding by binding to the 70 S ribosomal subunit.
  • Good inhibitory activity against M. tuberculosis (sensitive and drug-resistant).
  • Good inhibitory activity against K. pneumonia.
  • Used in studies of tuberculosis.
  • Used in studies of pneumonia.
Catalog No. 50-003-91271 Supplier Medchemexpress LLC Supplier No. HY16566A1G
Kanamycin (Kanamycin A) sulfate is an orally active antibacterial agent effective against gram-negative and gram-positive bacteria. It inhibits translocation and causes misencoding by binding to the 70 S ribosomal subunit. It shows good inhibitory activity against both M. tuberculosis (sensitive and drug-resistant) and K. pneumonia, and is used in studies of tuberculosis and pneumonia.
  • Orally active antibacterial agent.
  • Effective against gram-negative bacteria.
  • Effective against gram-positive bacteria.
  • Inhibits translocation.
  • Causes misencoding by binding to the 70 S ribosomal subunit.
  • Good inhibitory activity against M. tuberculosis (sensitive and drug-resistant).
  • Good inhibitory activity against K. pneumonia.
  • Used in studies of tuberculosis.
  • Used in studies of pneumonia.