Learn More
Medchemexpress LLC Kanamycin sulfate | 25389-94-0 | 100.0% | 582.58 | 100 G
SDP
Catalog No. 5000393836
Encompass_Preferred
Kanamycin sulfate (Kanamycin A sulfate) is an orally active antibacterial agent that targets both gram-negative and gram-positive bacteria. It works by inhibiting translocation and causing misencoding through binding to the 70S ribosomal subunit. It demonstrates good inhibitory activity against M. tuberculosis (both sensitive and drug-resistant strains) and K. pneumonia, making it suitable for studies related to tuberculosis and pneumonia.
  • Orally active antibacterial agent.
  • Effective against gram-negative and gram-positive bacteria.
  • Inhibits translocation and causes misencoding by binding to the 70S ribosomal subunit.
  • Good inhibitory activity against M. tuberculosis (sensitive and drug-resistant) and K. pneumonia.
  • Used in studies of tuberculosis and pneumonia.
Catalog No. 50-003-93836 Supplier Medchemexpress LLC Supplier No. HY16566A100G
Kanamycin sulfate (Kanamycin A sulfate) is an orally active antibacterial agent that targets both gram-negative and gram-positive bacteria. It works by inhibiting translocation and causing misencoding through binding to the 70S ribosomal subunit. It demonstrates good inhibitory activity against M. tuberculosis (both sensitive and drug-resistant strains) and K. pneumonia, making it suitable for studies related to tuberculosis and pneumonia.
  • Orally active antibacterial agent.
  • Effective against gram-negative and gram-positive bacteria.
  • Inhibits translocation and causes misencoding by binding to the 70S ribosomal subunit.
  • Good inhibitory activity against M. tuberculosis (sensitive and drug-resistant) and K. pneumonia.
  • Used in studies of tuberculosis and pneumonia.