Learn More
Medchemexpress LLC Kanamycin sulfate | 25389-94-0 | 99.9% | 582.58 | 25 G
SDP
Catalog No. 5000392581
Encompass_Preferred
Kanamycin sulfate is an orally active broad-spectrum antibacterial agent that inhibits translocation and causes misencoding by binding to the 70S ribosomal subunit. It shows good inhibitory activity against both gram-negative and gram-positive bacteria, including M. tuberculosis and K. pneumonia.
  • Effective against gram-negative and gram-positive bacteria.
  • Inhibits translocation by binding to the 70S ribosomal subunit.
  • Shows good inhibitory activity against sensitive and drug-resistant M. tuberculosis.
  • Exhibits good inhibitory activity against K. pneumonia.
  • Suitable for studies on tuberculosis and pneumonia.
  • Demonstrates in vitro activity against various mycobacteria strains (MICs of 1-5 μg/mL).
  • In vivo, inhibits growth of bovine tubercle bacilli in mice.
  • In vivo, inhibits multiplication of K. pneumonia DT-S in mice, increasing survival rate.
Catalog No. 50-003-92581 Supplier Medchemexpress LLC Supplier No. 25GHY16566A
Kanamycin sulfate is an orally active broad-spectrum antibacterial agent that inhibits translocation and causes misencoding by binding to the 70S ribosomal subunit. It shows good inhibitory activity against both gram-negative and gram-positive bacteria, including M. tuberculosis and K. pneumonia.
  • Effective against gram-negative and gram-positive bacteria.
  • Inhibits translocation by binding to the 70S ribosomal subunit.
  • Shows good inhibitory activity against sensitive and drug-resistant M. tuberculosis.
  • Exhibits good inhibitory activity against K. pneumonia.
  • Suitable for studies on tuberculosis and pneumonia.
  • Demonstrates in vitro activity against various mycobacteria strains (MICs of 1-5 μg/mL).
  • In vivo, inhibits growth of bovine tubercle bacilli in mice.
  • In vivo, inhibits multiplication of K. pneumonia DT-S in mice, increasing survival rate.