Learn More
Medchemexpress LLC Kanamycin sulfate | 25389-94-0 | 99.96% | 582.58 | 1 ML
SDP
Catalog No. 5000391795
Encompass_Preferred
Kanamycin sulfate is an orally active antibacterial agent effective against gram-negative and gram-positive bacteria. It inhibits translocation and causes misencoding by binding to the 70S ribosomal subunit, and demonstrates good inhibitory activity against M. tuberculosis and K. pneumonia.
  • Orally active antibacterial agent
  • Effective against gram-negative and gram-positive bacteria
  • Inhibits translocation by binding to the 70S ribosomal subunit
  • Causes misencoding by binding to the 70S ribosomal subunit
  • Good inhibitory activity against M. tuberculosis (sensitive and drug-resistant)
  • Good inhibitory activity against K. pneumonia
  • Suitable for studies of tuberculosis and pneumonia
Catalog No. 50-003-91795 Supplier Medchemexpress LLC Supplier No. HY16566A10MM/1ML
Kanamycin sulfate is an orally active antibacterial agent effective against gram-negative and gram-positive bacteria. It inhibits translocation and causes misencoding by binding to the 70S ribosomal subunit, and demonstrates good inhibitory activity against M. tuberculosis and K. pneumonia.
  • Orally active antibacterial agent
  • Effective against gram-negative and gram-positive bacteria
  • Inhibits translocation by binding to the 70S ribosomal subunit
  • Causes misencoding by binding to the 70S ribosomal subunit
  • Good inhibitory activity against M. tuberculosis (sensitive and drug-resistant)
  • Good inhibitory activity against K. pneumonia
  • Suitable for studies of tuberculosis and pneumonia