Learn More
Ubpbio Kanamycin Sulfate, 25G
Catalog No. 501965187
Encompass_Preferred
Kanamycin sulfate is an aminoglycosidic antibiotic that inhibits the growth of Gram-positive and Gram-negative bacteria and mycoplasmas in cell culture.
Catalog No. 50-196-5187 Supplier Ubpbio Supplier No. P801025
Kanamycin sulfate is an aminoglycosidic antibiotic that inhibits the growth of Gram-positive and Gram-negative bacteria and mycoplasmas in cell culture.