Learn More
Zymo Research Corporation Kanamycin Sulfate Solution, 5x5 ml
SDP
Catalog No. 502036986
Encompass
Kanamycin is effective against both Gram (-) and Gram (+) bacteria. Resistance to kanamycin is conferred by an aminoglycoside phosphotransferase that modifies the antibiotic, preventing its interaction with ribosomes. Kanamycin is commonly used to select for cosmid vectors.
Catalog No. 50-203-6986 Supplier Zymo Research Corporation Supplier No. A100325
Kanamycin is effective against both Gram (-) and Gram (+) bacteria. Resistance to kanamycin is conferred by an aminoglycoside phosphotransferase that modifies the antibiotic, preventing its interaction with ribosomes. Kanamycin is commonly used to select for cosmid vectors.