Learn More
Zymo Research Corporation Kanamycin Sulfate Solution, 1x5 ml
SDP
Catalog No. 501957628
Encompass
Kanamycin is effective against both Gram (-) and Gram (+) bacteria. Resistance to kanamycin is conferred by an aminoglycoside phosphotransferase that modifies the antibiotic, preventing its interaction with ribosomes. Kanamycin is commonly used to select for cosmid vectors.
Catalog No. 50-195-7628 Supplier Zymo Research Corporation Supplier No. A10035
Kanamycin is effective against both Gram (-) and Gram (+) bacteria. Resistance to kanamycin is conferred by an aminoglycoside phosphotransferase that modifies the antibiotic, preventing its interaction with ribosomes. Kanamycin is commonly used to select for cosmid vectors.