Learn More
Enzo Life Sciences Kanamycin . sulfate (5g). CAS: 25389-94-0
SDP
Catalog No. 502008932
Encompass_Preferred
Aminoglycoside antibiotic with broad antibacterial spectrum, including mycobacteria, many Gram-positive and most Gram-negative bacteria. Inhibits bacterial protein synthesis. Kanamycin resistance is widely used as a selection marker for obtaining transgenic plants. Solubility: Soluble in water. Long Term Storage: +4°C.
Catalog No. 50-200-8932 Supplier Enzo Life Sciences Supplier No. ALX380049G005
Aminoglycoside antibiotic with broad antibacterial spectrum, including mycobacteria, many Gram-positive and most Gram-negative bacteria. Inhibits bacterial protein synthesis. Kanamycin resistance is widely used as a selection marker for obtaining transgenic plants. Solubility: Soluble in water. Long Term Storage: +4°C.