Learn More
Medchemexpress LLC Kanamycin sulfate | 70560-51-9 | 99.0% | 582.58 g/mol | C18H38N4O15S | 5 G
SDP
Catalog No. 5000299527
Encompass_Preferred
Kanamycin sulfate is an aminoglycoside broad-spectrum antibiotic supplied as a research-grade powder used in microbiology and infection studies. It acts by inhibiting bacterial protein synthesis and is effective against many Gram-negative organisms and certain staphylococci; it is associated with ototoxicity and should be handled accordingly.
  • Broad-spectrum aminoglycoside antibiotic for research use.
  • Useful in studies of Gram-negative and certain staphylococcal infections.
  • Inhibits bacterial protein synthesis (bactericidal activity).
  • Available as a powder with high purity suitable for assays.
  • Soluble in water (62.5 mg/mL with ultrasonic agitation).
  • Requires storage at 4 °C, protected from moisture.
  • Associated with ototoxicity; handle with appropriate safety precautions.
Catalog No. 50-002-99527 Supplier Medchemexpress LLC Supplier No. HYW0401285G
Kanamycin sulfate is an aminoglycoside broad-spectrum antibiotic supplied as a research-grade powder used in microbiology and infection studies. It acts by inhibiting bacterial protein synthesis and is effective against many Gram-negative organisms and certain staphylococci; it is associated with ototoxicity and should be handled accordingly.
  • Broad-spectrum aminoglycoside antibiotic for research use.
  • Useful in studies of Gram-negative and certain staphylococcal infections.
  • Inhibits bacterial protein synthesis (bactericidal activity).
  • Available as a powder with high purity suitable for assays.
  • Soluble in water (62.5 mg/mL with ultrasonic agitation).
  • Requires storage at 4 °C, protected from moisture.
  • Associated with ototoxicity; handle with appropriate safety precautions.