Learn More
Abcam Kanamycin sulfate (aqueous), aminoglycoside antibiotic agent, 10ML
Catalog No. 502704567
Encompass_Preferred
MW 582.6 Da. Aminoglycoside antibiotic agent effective against Gram-negative and Gram-positive bacteria and mycoplasma. Originally purified from the bacterium Streptomyces kanamyceticus. Binds to the bacterial 30S ribosomal subunit, inhibits translocation, and elicits miscoding. Often used to select for bacteria which have been successfully transformed with a plasmid conferring kanamycin resistance.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4567 Supplier Abcam Supplier No. AB14657410ML
MW 582.6 Da. Aminoglycoside antibiotic agent effective against Gram-negative and Gram-positive bacteria and mycoplasma. Originally purified from the bacterium Streptomyces kanamyceticus. Binds to the bacterial 30S ribosomal subunit, inhibits translocation, and elicits miscoding. Often used to select for bacteria which have been successfully transformed with a plasmid conferring kanamycin resistance.
The product is subject to the following: Abcam Restricted Use Statement