Learn More
Crescent Chemical Co Inc KANAMYCIN SULFATE MOLECULAR
Supplier: Crescent Chemical Co Inc SE26899.02
Kanamycin sulfate molecular biology grade, Ph. Eur. Aminoglycoside antibiotic, inhibitor of protein biosynthesis. It is active against gram negative and gram positive bacteria. Main component is Kanamycin A. Suitable for prevention of bacterial contamination in cell