Learn More
Crescent Chemical Co Inc KANAMYCIN SULFATE MOLECULAR
SDP
Catalog No. 50247775
Encompass_Preferred
Kanamycin sulfate molecular biology grade, Ph. Eur. Aminoglycoside antibiotic, inhibitor of protein biosynthesis. It is active against gram negative and gram positive bacteria. Main component is Kanamycin A. Suitable for prevention of bacterial contamination in cell
Catalog No. 50-247-775 Supplier Crescent Chemical Co Inc Supplier No. SE26899.03
Kanamycin sulfate molecular biology grade, Ph. Eur. Aminoglycoside antibiotic, inhibitor of protein biosynthesis. It is active against gram negative and gram positive bacteria. Main component is Kanamycin A. Suitable for prevention of bacterial contamination in cell