Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kanamycin Sulfate Solution, 50 mg/mL, USP Grade, G-Biosciences
SCI_ED

Catalog No. S139717
Change view
Click to view available options
Quantity:
25 mL
50 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S139717 50 mL
S139844 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S139717 Supplier G-Biosciences Supplier No. RC1044
Add to Cart
Add to Cart

Kanamycin is an aminoglycoside antibiotic and has a bactericidal action against many gram-negative bacteria.

Kanamycin is used as a selective agent for the incorporation of the NPT II (APH3) gene in plant tissues.

Specifications

Type α-Carboxybenzyl Penicillin Disodium Salt
Physical Form Suspension
CAS 25389-94-0
Sterility Non-sterile
Color White
Concentration 50 mg/mL
Quantity 50 mL
Percent Purity 99%
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.