Learn More
Medchemexpress LLC Kanamycins sulfate | 70560-51-9 | 99.0% | 582.58 g/mol | C18H38N4O15S | 1 G
SDP
Catalog No. 5000301977
Encompass_Preferred
Kanamycins sulfate is an aminoglycoside, broad-spectrum antibiotic provided as the sulfate salt for laboratory and research use. It is active against certain severe staphylococcal and Gram-negative bacillary infections and should be handled according to safety guidance due to known ototoxicity.
  • Broad-spectrum aminoglycoside antibiotic suitable for research applications.
  • Active against certain severe staphylococcal and Gram-negative bacillary infections.
  • Provided as the sulfate salt for consistent solubility and formulation.
  • High reported purity, supporting experimental reproducibility.
  • Known ototoxicity; use appropriate safety precautions and handling procedures.
Catalog No. 50-003-01977 Supplier Medchemexpress LLC Supplier No. HYW0401281G
Kanamycins sulfate is an aminoglycoside, broad-spectrum antibiotic provided as the sulfate salt for laboratory and research use. It is active against certain severe staphylococcal and Gram-negative bacillary infections and should be handled according to safety guidance due to known ototoxicity.
  • Broad-spectrum aminoglycoside antibiotic suitable for research applications.
  • Active against certain severe staphylococcal and Gram-negative bacillary infections.
  • Provided as the sulfate salt for consistent solubility and formulation.
  • High reported purity, supporting experimental reproducibility.
  • Known ototoxicity; use appropriate safety precautions and handling procedures.