Learn More
Honeywell™ Kangaroo Leather Super Glove™ - Knit Wrist
Catalog No. 19171020B
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-171-020B Small
19-171-020C Medium
19-171-020D Large
19-171-020E X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-171-020B Supplier Honeywell™ Supplier No. GLSGKCW S
Add to Cart
Add to Cart
3D hand-shaped design with staggered layer seaming is ergonomically correct, helping to fight hand fatigue and reduce injuries. Digiroo™ High Grip technology on the palm and fingers ensures dexterity, grip, and tactility.
  • Air Spacer™ Thermal Architecture traps air for thermal protection without adding bulk to the glove
  • Black Digiroo™ Digital Kangaroo Grip on the palm and fingers for durability, grip, and tactility
  • Crosstech™ Moisture Barrier is stable and durable with high temperature resistance
  • Bubble-Flex Construction on the knuckles and fingers features dead-air spacer ridges for natural hand flexing
  • Top Grain Kangaroo Leather provides abrasion resistance and exceptional durability
  • Anthropomorphically correct 3D-style fights hand fatigue and reduces injury
Certifications
NFPA 1971-2007, NFPA 1951, NFPA 1999, NFPA 1992
TRUSTED_SUSTAINABILITY
Specifications
Back Material Top Grain Kangaroo Leather
Certifications/Compliance NFPA 1971-2007, NFPA 1951, NFPA 1999, NFPA 1992
Coated Area Palm, Fingers
Cuff Material Nomex™
Color Blue
Hand Specific
Product Type Structural Firefighting Gloves
Size Small
Glove Cuff Color Black
Glove Cuff Style Knit
Includes Crosstech™ Moisture Barrier
Liner Material Nomex™/Aramid Fiber
Series GL-SGKCW
Thumb Style Wing