Learn More
Description
- Can you Kanoodle? Using combinations of colored connected beads, players complete colorful logic puzzles in two unique formats: tricky 2D puzzles and twisted 3D pyramids.
- To start, pick a design from the puzzle book, place some of the puzzle pieces into position as shown…then fill the empty spaces with the remaining pieces.
- Sounds simple? It can be…or it can be deviously difficult, depending on the level of game play. Hundreds of possible combinations, but only one completes the puzzle.
- Puzzle pieces fit inside the handy pocket-sized case with built-in game boards, so you can Kanoodle anywhere.
- Includes 12 puzzle pieces, 48-page illustrated puzzle book, and carrying case.
- For 1 player.
Specifications
Specifications
| Grade Level | Elementary, Intermediate - Middle School, High School |
| Grade | K to 12 |
| Class Size | 1 Student |
| Product Type | 2D/3D Puzzle |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
