Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Kanosamine hydrochloride | 57649-10-2 | MFCD01725274 | ≥98.0% | 215.63 g/mol | C6H14ClNO5 | 5 MG
SDP

Catalog No. 5000264164
Encompass_Preferred

Kanosamine hydrochloride is the hydrochloride salt of kanosamine, an aminoglycoside sugar antibiotic used in biochemical and microbiological research. It inhibits certain plant-pathogenic oomycetes, select fungi, and some bacteria and is employed as a reference compound to study cell wall biosynthesis and pathogen growth.

  • Hydrochloride salt form of kanosamine.
  • CAS 57649-10-2.
  • Molecular formula C6H14ClNO5; molecular weight 215.63 g/mol.
  • Reported activity: inhibits Phytophthora medicaginis and Aphanomyces euteiches (MICs 25 and 60 μg/mL).
  • Available in small research quantities (for example, 5 mg).
  • Purity commonly ≥98% depending on supplier and lot.
  • Used as a reference standard in microbiology and biochemical studies.

Catalog No. 50-002-64164 Supplier Medchemexpress LLC Supplier No. HY1121765MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Kanosamine hydrochloride is the hydrochloride salt of kanosamine, an aminoglycoside sugar antibiotic used in biochemical and microbiological research. It inhibits certain plant-pathogenic oomycetes, select fungi, and some bacteria and is employed as a reference compound to study cell wall biosynthesis and pathogen growth.

  • Hydrochloride salt form of kanosamine.
  • CAS 57649-10-2.
  • Molecular formula C6H14ClNO5; molecular weight 215.63 g/mol.
  • Reported activity: inhibits Phytophthora medicaginis and Aphanomyces euteiches (MICs 25 and 60 μg/mL).
  • Available in small research quantities (for example, 5 mg).
  • Purity commonly ≥98% depending on supplier and lot.
  • Used as a reference standard in microbiology and biochemical studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.